Assay Details
| Gene Symbol: Atad3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_179203.3 | NP_849534.2 | 10 - 11 | 1182 | 93 | |
| GenBank mRNA | AF343079.1 | - | 10 - 11 | 1228 | 93 | |
| AK075921.1 | - | 10 - 11 | 1178 | 93 | ||
| AK148974.1 | - | 10 - 11 | 1181 | 93 | ||
| AK149702.1 | - | 10 - 11 | 1181 | 93 | ||
| AK150037.1 | - | 10 - 11 | 1181 | 93 | ||
| AK166099.1 | - | 10 - 11 | 1178 | 93 | ||
| AK172038.1 | - | 10 - 11 | 1178 | 93 | ||
| AK172350.1 | - | 10 - 11 | 1183 | 93 | ||
| AK173141.1 | - | 10 - 11 | 1130 | 93 | ||
| BC023301.1 | - | 10 - 11 | 1119 | 3671787 | 93 | |
| BC058373.1 | - | 10 - 11 | 1166 | 6828037 | 93 | |
| BC060036.1 | - | 10 - 11 | 1114 | 6530155 | 93 | |
More Information
Set Membership: |
|

Atad3a
Genomic Map