Assay Details
| Gene Symbol: Magoh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001282737.1 | NP_001269666.1 | 3 - 4 | 360 | 119 | |
| GenBank mRNA | AF007862.1 | - | 3 - 4 | 347 | 119 | |
| AF035939.1 | - | 3 - 4 | 307 | 119 | ||
| AK011512.1 | - | 3 - 4 | 299 | 119 | ||
| BC018176.1 | - | 3 - 4 | 298 | 3587774 | 119 | |
More Information
Updated Mapping Information
Set Membership: |
|

Magoh
Genomic Map