Assay Details
| Gene Symbol: Wdr31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290522.1 | NP_001277451.1 | 3 - 4 | 324 | 72 | |
| GenBank mRNA | AK017259.1 | - | 3 - 4 | 317 | 72 | |
More Information
Set Membership: |
|

Wdr31
Genomic Map