Assay Details
| Gene Symbol: Pole3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021498.2 | NP_067473.2 | 1 - 2 | 86 | 62 | |
| GenBank mRNA | AF230806.1 | - | 1 - 2 | 86 | 62 | |
| AK007693.1 | - | 141 | 62 | |||
| BC024996.1 | - | 1 - 2 | 86 | 5253060 | 62 | |
| BC034004.1 | - | 94 | 62 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Pole3
Genomic Map