| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0100004701.mm.1 |
NM_001271533.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100004701.mm.1 |
NM_029160.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100004701.mm.1 |
TC0100000585.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100004702.mm.1 |
NM_001271533.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100004702.mm.1 |
NM_029160.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100004702.mm.1 |
TC0100000585.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100004699.mm.1 |
NM_001271533.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100004699.mm.1 |
NM_029160.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100004699.mm.1 |
TC0100000585.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100004700.mm.1 |
NM_001271533.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100004700.mm.1 |
NM_029160.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100004700.mm.1 |
TC0100000585.mm.1 |
MTA TC |