Assay Details
| Gene Symbol: Fam134a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK165957.1 | - | 9 - 10 | 1347 | 112 | |
| AK210282.1 | - | 2 - 3 | 344 | 112 | ||
| AK210397.1 | - | 2 - 3 | 344 | 112 | ||
| BC026519.1 | - | 5 - 6 | 719 | 112 | ||
| BC038286.1 | - | 5 - 6 | 754 | 4221441 | 112 | |
| BC070406.1 | - | 9 - 10 | 1360 | 5707319 | 112 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fam134a
Genomic Map