Assay Details
| Gene Symbol: Fut8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252614.1 | NP_001239543.1 | 1 - 2 | 94 | 75 | |
| NM_016893.5 | NP_058589.2 | 1 - 2 | 94 | 75 | ||
| GenBank mRNA | AK051811.1 | - | 1 - 2 | 51 | 75 | |
| BC010666.1 | - | 1 - 2 | 73 | 3594582 | 75 | |
| BU702757.1 | - | 1 - 2 | 125 | 75 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK051811.1, BC010666.1, NM_001252614.1, NM_016893.5 transcripts.
Set Membership: |
|

Fut8
Genomic Map