Assay Details
| Gene Symbol: Kansl1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001122738.1 | NP_001116210.1 | 12 - 13 | 2611 | 105 | |
| NM_177645.4 | NP_808313.3 | 11 - 12 | 2584 | 105 | ||
| GenBank mRNA | AK082897.1 | - | 11 - 12 | 1431 | 105 | |
| AK220534.1 | - | 11 - 12 | 2535 | 105 | ||
| BC141036.1 | - | 12 - 13 | 2494 | 9055648 | 105 | |
More Information
Updated Mapping Information
Set Membership: |
|

Kansl1l
Genomic Map