Assay Details
| Gene Symbol: Kansl1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001122738.1 | NP_001116210.1 | 4 - 5 | 1579 | 103 | |
| NM_177645.4 | NP_808313.3 | 4 - 5 | 1684 | 103 | ||
| GenBank mRNA | AK082897.1 | - | 3 - 4 | 399 | 103 | |
| AK145440.1 | - | 4 - 5 | 1656 | 103 | ||
| AK220534.1 | - | 4 - 5 | 1635 | 103 | ||
| BC141036.1 | - | 5 - 6 | 1594 | 9055648 | 103 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Kansl1l
Genomic Map