Assay Details
| Gene Symbol: Gm15179 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_037976.1 | 1 - 2 | 130 | 88 | ||
| GenBank mRNA | AK039780.1 | - | 1 - 2 | 128 | 88 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | desmin | Des | -4.8kb | Anti-sense | Chr. 1 - 75360292 - 75368579 on NCBI Build GRCm38 | |
| Downstream | SPEG complex locus | Speg |
AW125581, Apeg1, BPEG, D1Bwg1450e, SPEGalpha, SPEGbeta, mKIAA1297 |
285.0 | Anti-sense | Chr. 1 - 75375297 - 75432320 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Gm15179
Genomic Map