Assay Details
| Gene Symbol: Plac8l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027072.1 | NP_081348.1 | 2 - 3 | 341 | 72 | |
| GenBank mRNA | AK006002.1 | - | 2 - 3 | 319 | 72 | |
| BC110558.1 | - | 2 - 3 | 303 | 40049443 | 72 | |
| BC110559.1 | - | 2 - 3 | 303 | 40049451 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|

Plac8l1
Genomic Map