Assay Details
| Gene Symbol: Spinkl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_183123.2 | NP_898946.1 | 3 - 4 | 162 | 87 | |
| GenBank mRNA | AK020535.1 | - | 3 - 4 | 163 | 87 | |
| EU234015.1 | - | 540 | 87 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Spinkl
Genomic Map