Assay Details
| Gene Symbol: Gm12802 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK084081.1 | - | 1 - 2 | 389 | 118 | |
| BB657329.1 | - | 1 - 2 | 389 | 118 | ||
| BG807815.1 | - | 1 - 2 | 174 | 118 | ||
| CX568958.1 | - | 1 - 2 | 345 | 119 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | CUB domain containing protein 2 | Cdcp2 | D030010E02Rik | -12.3kb | Sense | Chr. 4 - 107096891 - 107114304 on NCBI Build GRCm38 |
| Downstream | transcription elongation factor A (SII) N-terminal and central domain containing 2 | Tceanc2 |
2210010B22Rik, 2210012G02Rik, 6330404A07Rik, AU015593, Tdeanc2 |
3.7kb | Anti-sense | Chr. 4 - 107134162 - 107178366 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm12802
Genomic Map