Assay Details
| Gene Symbol: Gm12802 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK084081.1 | - | 2 - 3 | 521 | 67 | |
| BB657329.1 | - | 2 - 3 | 521 | 67 | ||
| BG807815.1 | - | 2 - 3 | 306 | 67 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | CUB domain containing protein 2 | Cdcp2 | D030010E02Rik | -16.0kb | Sense | Chr. 4 - 107096891 - 107114304 on NCBI Build GRCm38 |
| Downstream | transmembrane protein 59 | Tmem59 |
1110001M20Rik, 3110046P06Rik, AI256529, D4Ertd20e, MTDCF1, ORF18 |
43.7kb | Sense | Chr. 4 - 107178630 - 107200996 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm12802
Genomic Map