Assay Details
| Gene Symbol: Sgpp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK087798.1 | - | 3 - 4 | 715 | 63 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | predicted gene 2102 | Gm2102 | -106.6kb | Sense | Chr. 1 - 78214966 - 78218981 on NCBI Build GRCm38 | |
| Downstream |
phenylalanyl-tRNA synthetase, beta subunit |
Farsb |
C76708, Farsa, Farsl, Farslb, Frsb |
90.3kb | Sense | Chr. 1 - 78417958 - 78488897 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Sgpp2
Genomic Map