Assay Details
| Gene Symbol: Des | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010043.2 | NP_034173.1 | 7 - 8 | 1414 | 73 | |
| GenBank mRNA | AK131922.1 | - | 7 - 8 | 1374 | 73 | |
| AK132391.1 | - | 7 - 8 | 1378 | 73 | ||
| BC031760.1 | - | 7 - 8 | 1362 | 4219280 | 73 | |
| L22550.1 | - | 7 - 8 | 1371 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Des
Genomic Map