Assay Details
| Gene Symbol: Gm26676 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK039137.1 | - | 1 - 2 | 32 | 79 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
potassium voltage-gated channel, subfamily G, member 2 |
Kcng2 | AA591524 | Anti-sense | Chr. 18 - 80294544 - 80365105 on NCBI Build GRCm38 | |
| Downstream |
CTD (carboxy-terminal domain, RNA polymerase II, polypeptide A) phosphatase, subunit 1 |
Ctdp1 |
4930563P03Rik, AW553592 |
83.6kb | Anti-sense | Chr. 18 - 80407959 - 80470612 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm26676
Genomic Map