Assay Details
Gene Symbol: 1700065O20Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_045386.1 | 3 - 4 | 403 | 70 | ||
GenBank mRNA | AK006898.1 | - | 3 - 4 | 406 | 70 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | predicted gene 16283 | Gm16283 | OTTMUSG00000033143 | -44.2kb | Anti-sense | Chr. 18 - 49755890 - 49761581 on NCBI Build GRCm38 |
Downstream | Dmx-like 1 | Dmxl1 |
C630007L23Rik, D030030C22 |
25.6kb | Anti-sense | Chr. 18 - 49832993 - 49965473 on NCBI Build GRCm38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
