Assay Details
| Gene Symbol: Gm13049 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK033051.1 | - | 460 | 66 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
solute carrier family 45, member 1 |
Slc45a1 |
C230078B22, Dnb5 |
-179.8kb | Anti-sense | Chr. 4 - 150629396 - 150652174 on NCBI Build GRCm38 |
| Downstream | ERBB receptor feedback inhibitor 1 | Errfi1 |
1300002F13Rik, AI788755, Mig-6, Mig6, RALT |
14.3kb | Sense | Chr. 4 - 150853801 - 150868889 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm13049
Genomic Map