Assay Details
| Gene Symbol: Gm12708 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK089375.1 | - | 2 - 3 | 188 | 73 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
jun proto-oncogene, opposite strand |
Junos | -37.2kb | Sense | Chr. 4 - 95052329 - 95169036 on NCBI Build GRCm38 | |
| Downstream | predicted gene 830 | Gm830 | 257.5kb | Anti-sense | Chr. 4 - 95466913 - 95487394 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Gm12708
Genomic Map