Assay Details
| Gene Symbol: Arhgef16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112744.1 | NP_001106215.1 | 4 - 5 | 909 | 106 | |
| GenBank mRNA | AK042195.1 | - | 4 - 5 | 905 | 106 | |
| AK153708.1 | - | 4 - 5 | 1070 | 106 | ||
| BC020030.1 | - | 1 - 2 | 70 | 106 | ||
| BC044805.1 | - | 4 - 5 | 1888 | 106 | ||
| BC095966.1 | - | 2 - 3 | 206 | 106 | ||
| BC137847.1 | - | 4 - 5 | 950 | 8860866 | 106 | |
| BC144970.1 | - | 4 - 5 | 950 | 9053518 | 106 | |
| BC144971.1 | - | 4 - 5 | 973 | 106 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Arhgef16
Genomic Map