Assay Details
| Gene Symbol: Arhgef16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112744.1 | NP_001106215.1 | 11 - 12 | 1723 | 72 | |
| GenBank mRNA | AK042195.1 | - | 11 - 12 | 1719 | 72 | |
| BC020030.1 | - | 8 - 9 | 884 | 72 | ||
| BC026661.1 | - | 3 - 4 | 736 | 72 | ||
| BC044805.1 | - | 10 - 11 | 3137 | 72 | ||
| BC095966.1 | - | 9 - 10 | 1020 | 72 | ||
| BC137847.1 | - | 11 - 12 | 1764 | 8860866 | 72 | |
| BC144970.1 | - | 11 - 12 | 1764 | 9053518 | 72 | |
| BC144971.1 | - | 11 - 12 | 1787 | 72 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Arhgef16
Genomic Map