Assay Details
| Gene Symbol: Arhgef16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112744.1 | NP_001106215.1 | 13 - 14 | 1985 | 62 | |
| GenBank mRNA | AK042195.1 | - | 13 - 14 | 1981 | 62 | |
| AK153708.1 | - | 13 - 14 | 2146 | 62 | ||
| BC020030.1 | - | 10 - 11 | 1146 | 62 | ||
| BC044805.1 | - | 12 - 13 | 3399 | 62 | ||
| BC095966.1 | - | 11 - 12 | 1282 | 62 | ||
| BC137847.1 | - | 13 - 14 | 2026 | 8860866 | 62 | |
| BC144970.1 | - | 13 - 14 | 2026 | 9053518 | 62 | |
| BC144971.1 | - | 13 - 14 | 2049 | 62 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Arhgef16
Genomic Map