Assay Details
| Gene Symbol: Rab27b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK049854.1 | - | 2 - 3 | 135 | 96 | |
| BY130221.1 | - | 2 - 3 | 139 | 96 | ||
| BY721478.1 | - | 2 - 3 | 133 | 96 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK049854.1 transcripts.
Set Membership: |
|

Rab27b
Genomic Map