Assay Details
| Gene Symbol: Acer2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290541.1 | NP_001277470.1 | 2 - 3 | 298 | 78 | |
| NM_001290543.1 | NP_001277472.1 | 2 - 3 | 298 | 78 | ||
| NM_139306.3 | NP_647467.1 | 2 - 3 | 298 | 78 | ||
| GenBank mRNA | AF282864.1 | - | 2 - 3 | 257 | 78 | |
| AK085306.1 | - | 2 - 3 | 294 | 78 | ||
| AY312515.1 | - | 2 - 3 | 223 | 78 | ||
| BC059819.1 | - | 2 - 3 | 298 | 6839525 | 78 | |
| CB723138.1 | - | 2 - 3 | 298 | 78 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Acer2
Genomic Map