Assay Details
| Gene Symbol: Acer2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290541.1 | NP_001277470.1 | 3 - 4 | 440 | 79 | |
| GenBank mRNA | BC059819.1 | - | 3 - 4 | 440 | 6839525 | 79 |
| CB723138.1 | - | 3 - 4 | 79 | |||
More Information
Set Membership: |
|

Acer2
Genomic Map