Assay Details
| Gene Symbol: Rbm20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001170847.1 | NP_001164318.1 | 10 - 11 | 2698 | 68 | |
| GenBank mRNA | BC038663.1 | - | 2 - 3 | 734 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rbm20
Genomic Map