Assay Details
| Gene Symbol: Pyroxd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_183165.3 | NP_898988.2 | 6 - 7 | 674 | 123 | |
| GenBank mRNA | AK148411.1 | - | 713 | 123 | ||
| AK165677.1 | - | 6 - 7 | 677 | 123 | ||
| BC022914.1 | - | 6 - 7 | 693 | 123 | ||
| BC027061.1 | - | 6 - 7 | 699 | 5355217 | 123 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pyroxd1
Genomic Map