Assay Details
| Gene Symbol: Grin2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008171.3 | NP_032197.3 | 1 - 2 | 88 | ||
| GenBank mRNA | AJ459261.1 | - | 1 - 2 | 88 | ||
| AK002963.1 | - | 1 - 2 | 88 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the AJ459261.1, AK002963.1, NM_008171.3 transcripts.
Set Membership: |
|

Grin2b
Genomic Map