Assay Details
| Gene Symbol: Grin2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008171.3 | NP_032197.3 | 3 - 4 | 743 | 62 | |
| GenBank mRNA | AJ459261.1 | - | 3 - 4 | 744 | 62 | |
| AJ459262.1 | - | 3 - 4 | 578 | 62 | ||
| AJ459263.1 | - | 2 - 3 | 505 | 62 | ||
| AK002963.1 | - | 3 - 4 | 744 | 62 | ||
| AK163195.1 | - | 473 | 62 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the AJ459261.1, AJ459262.1, AJ459263.1, AK002963.1, AK163195.1, NM_008171.3 transcripts.
Set Membership: |
|

Grin2b
Genomic Map