| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1200003208.mm.1 |
NM_025373.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200003208.mm.1 |
NM_025373.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200003208.mm.1 |
TC1200000464.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1200003212.mm.1 |
NM_025373.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200003212.mm.1 |
NM_025373.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200003212.mm.1 |
TC1200000464.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1200003213.mm.1 |
NM_025373.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200003213.mm.1 |
NM_025373.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200003213.mm.1 |
TC1200000464.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1200003211.mm.1 |
NM_025373.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200003211.mm.1 |
NM_025373.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200003211.mm.1 |
TC1200000464.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1200003209.mm.1 |
NM_025373.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200003209.mm.1 |
NM_025373.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200003209.mm.1 |
TC1200000464.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1200003214.mm.1 |
NM_025373.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200003214.mm.1 |
NM_025373.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200003214.mm.1 |
TC1200000464.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1200003205.mm.1 |
NM_025373.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200003205.mm.1 |
NM_025373.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200003205.mm.1 |
TC1200000464.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1200003206.mm.1 |
NM_025373.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200003206.mm.1 |
NM_025373.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200003206.mm.1 |
TC1200000464.mm.1 |
MTA TC |