Assay Details
| Gene Symbol: Riok3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024182.4 | NP_077144.2 | 4 - 5 | 493 | 90 | |
| GenBank mRNA | AK131635.1 | - | 4 - 5 | 497 | 90 | |
| AK134327.1 | - | 4 - 5 | 585 | 90 | ||
| AK147074.1 | - | 4 - 5 | 494 | 90 | ||
| AK167309.1 | - | 4 - 5 | 589 | 90 | ||
| AK170882.1 | - | 4 - 5 | 573 | 90 | ||
| BC033271.1 | - | 4 - 5 | 531 | 4016197 | 90 | |
More Information
Updated Mapping Information
Set Membership: |
|

Riok3
Genomic Map