Assay Details
| Gene Symbol: Riok3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024182.4 | NP_077144.2 | 9 - 10 | 1239 | 123 | |
| GenBank mRNA | AK004748.1 | - | 9 - 10 | 1331 | 123 | |
| AK011110.1 | - | 2 - 3 | 323 | 123 | ||
| AK131635.1 | - | 9 - 10 | 1243 | 123 | ||
| AK134327.1 | - | 9 - 10 | 1331 | 123 | ||
| AK147074.1 | - | 9 - 10 | 1240 | 123 | ||
| AK167309.1 | - | 9 - 10 | 1335 | 123 | ||
| AK170882.1 | - | 9 - 10 | 1319 | 123 | ||
| BC002255.1 | - | 6 - 7 | 769 | 123 | ||
| BC033271.1 | - | 9 - 10 | 1277 | 4016197 | 123 | |
More Information
Updated Mapping Information
Set Membership: |
|

Riok3
Genomic Map