Assay Details
| Gene Symbol: Rbbp8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081223.2 | NP_001074692.1 | 15 - 16 | 2712 | 70 | |
| NM_001252495.1 | NP_001239424.1 | 15 - 16 | 2617 | 70 | ||
| NR_045526.1 | 14 - 15 | 2585 | 70 | |||
| NR_045527.1 | 14 - 15 | 2556 | 70 | |||
| GenBank mRNA | AK037049.1 | - | 15 - 16 | 2635 | 70 | |
| AK041384.1 | - | 14 - 15 | 2462 | 70 | ||
| AK077614.1 | - | 14 - 15 | 2664 | 70 | ||
| AK163908.1 | - | 2552 | 70 | |||
| BC050849.1 | - | 15 - 16 | 2617 | 6508067 | 70 | |
| CX567593.1 | - | 1 - 2 | 149 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rbbp8
Genomic Map