Assay Details
Gene Symbol: Gm553 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC051519.1 | - | 348 | 73 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Downstream | transmembrane protein 194B | Tmem194b |
5330401P04Rik, 9830163O08 |
15.9kb | Sense | Chr. 1 - 52568535 - 52651919 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
