Assay Details
| Gene Symbol: Gm553 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC051519.1 | - | 400 | 62 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | predicted gene 28178 | Gm28178 | Anti-sense | Chr. 1 - 52570047 - 52580292 on NCBI Build GRCm38 | ||
| Downstream | major facilitator superfamily domain containing 6 | Mfsd6 |
2210010L05Rik, 9630025I22Rik, AI836200, AW212394, MMR2 |
86.2kb | Anti-sense | Chr. 1 - 52656299 - 52727462 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm553
Genomic Map