Assay Details
| Gene Symbol: Carf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285463.1 | NP_001272392.1 | 15 - 16 | 1827 | 74 | |
| NM_001285473.1 | NP_001272402.1 | 11 - 12 | 1417 | 74 | ||
| NM_139150.5 | NP_631889.1 | 13 - 14 | 1700 | 74 | ||
| NR_104344.1 | 9 - 10 | 1152 | 74 | |||
| GenBank mRNA | AB221600.1 | - | 12 - 13 | 1483 | 74 | |
| AF454947.1 | - | 13 - 14 | 1669 | 74 | ||
| AK044738.1 | - | 15 - 16 | 1828 | 74 | ||
| BC138644.1 | - | 12 - 13 | 1606 | 8861664 | 74 | |
| BC138645.1 | - | 12 - 13 | 1606 | 8861665 | 74 | |
| BC145350.1 | - | 12 - 13 | 1596 | 74 | ||
| EU170502.1 | - | 11 - 12 | 1398 | 74 | ||
| EU170503.1 | - | 9 - 10 | 1133 | 74 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Carf
Genomic Map