Assay Details
| Gene Symbol: Carf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285463.1 | NP_001272392.1 | 4 - 5 | 462 | 72 | |
| NM_001285473.1 | NP_001272402.1 | 1 - 2 | 119 | 72 | ||
| NR_104344.1 | 1 - 2 | 119 | 72 | |||
| GenBank mRNA | AK044738.1 | - | 4 - 5 | 463 | 72 | |
| EU170502.1 | - | 1 - 2 | 100 | 72 | ||
| EU170503.1 | - | 1 - 2 | 100 | 72 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001285463.1 transcripts.
Set Membership: |
|

Carf
Genomic Map