Assay Details
| Gene Symbol: 4933424G05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045372.1 | 4 - 5 | 480 | 74 | ||
| GenBank mRNA | AK016892.1 | - | 4 - 5 | 481 | 74 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | potassium channel tetramerisation domain containing 1 | Kctd1 |
4933402K10Rik, AI661543, AW553000 |
Anti-sense | Chr. 18 - 14968685 - 15151446 on NCBI Build GRCm38 | |
| Downstream |
aldehyde dehydrogenase 9, subfamily A1 pseudogene |
Gm2629 | 165.1kb | Sense | Chr. 18 - 15213100 - 15214264 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

4933424G05Rik
Genomic Map