Assay Details
| Gene Symbol: Gm12709 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_040444.1 | 1 - 2 | 206 | 71 | ||
| GenBank mRNA | AK038336.1 | - | 1 - 2 | 207 | 71 | |
| BC064118.1 | - | 1 - 2 | 172 | 71 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | SH3-domain GRB2-like (endophilin) interacting protein 1 | Sgip1 |
3110007P09Rik, SGIP1alpha, gag |
-11.2kb | Anti-sense | Chr. 4 - 102759167 - 102972701 on NCBI Build GRCm38 |
| Downstream | expressed sequence BB031773 | BB031773 | 14.4kb | Sense | Chr. 4 - 103004106 - 103011965 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Gm12709
Genomic Map