Assay Details
| Gene Symbol: C87499 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198663.3 | NP_941065.2 | 3 - 4 | 954 | 137 | |
| NR_027385.1 | 2 - 3 | 643 | 137 | |||
| GenBank mRNA | AK135883.1 | - | 3 - 4 | 954 | 137 | |
| AK163216.1 | - | 2 - 3 | 645 | 137 | ||
| BC053719.1 | - | 3 - 4 | 955 | 30053512 | 137 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2028675 | AK135883.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2028675 | BC053719.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2028675 | CA559642.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2028675 | NM_198663.3 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 198194 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 450511 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 439634 | AK029877.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 439634 | BC145660.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 439634 | NM_207268.3 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 2602 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 132100 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P141931 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2028675 | AK135883.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2028675 | BC053719.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2028675 | CA559642.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2028675 | NM_198663.3 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23091474 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23091509 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23091508 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23091486 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23091492 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23091468 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23091506 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23091511 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23091478 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23091483 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23091490 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23091467 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23091477 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23091514 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23091480 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P141931 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P00924434 | - | - |
| Whole Mouse Genome G4122A | A_52_P187183 | AK163216.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P187183 | AK135883.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P187183 | BC053719.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P187183 | NM_198663.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P187183 | NR_027385.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P130047 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P141931 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03687802 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03687805 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03687796 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03687801 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03687804 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03687799 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03687798 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23091429 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23091454 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23091447 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23091500 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23091438 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23091492 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23091426 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23091478 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23091423 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23091486 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23091508 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23091464 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23091506 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23091448 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23091435 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23091443 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23091511 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23091474 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23091496 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23091433 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23091442 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23091477 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23091494 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23091509 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23091451 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23091458 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23091514 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23091502 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23091425 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23091441 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23091508 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23091443 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23091511 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03687798 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23091481 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23091506 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23091493 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03912758 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03687805 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03517553 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03554103 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23091490 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03630716 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23091500 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04076526 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23091469 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23091465 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03630719 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23091474 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23091433 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03630710 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23091492 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23091514 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04076522 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23091483 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03687801 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23091429 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04076525 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03425769 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03653589 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03258212 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03997735 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04064151 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23091451 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23091504 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03912763 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03630717 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23091467 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23091426 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23091442 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23091477 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03912764 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03912757 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04076517 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23091447 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23091480 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23091448 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23091484 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23091509 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10514307 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10514308 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10514305 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10514306 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1460611_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10514304 | - | - |
| Mouse Genome 430 2.0 Array | 1460611_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1460611_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 381590_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 381590_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0400024311.mm.1 | TC0400003008.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400024311.mm.1 | NM_198663.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400024311.mm.1 | NM_198663.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400024311.mm.1 | NR_027385.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400024311.mm.1 | NR_027385.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400024313.mm.1 | TC0400003008.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400024313.mm.1 | NM_198663.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400024313.mm.1 | NM_198663.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400024313.mm.1 | NR_027385.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400024313.mm.1 | NR_027385.1 | Refseq |
| Mouse Expression Set 430 | 1450593_PM_at | - | - |
| Mouse Expression Set 430 | 1422403_at | - | - |
| Mouse Expression Set 430 | 1422406_PM_at | - | - |
| Mouse Expression Set 430 | 1422403_PM_at | - | - |
| Mouse Expression Set 430 | 1450593_at | - | - |
| Mouse Expression Set 430 | 1422404_PM_x_at | - | - |
| Mouse Expression Set 430 | 1422404_x_at | - | - |
| Mouse Expression Set 430 | 1422406_at | - | - |
| Mouse Expression Set 430 | 1460611_PM_at | - | - |
| Mouse Expression Set 430 | 1460611_at | - | - |
| Murine Genome U74v2 Set | 101619_at | - | - |
| Murine Genome U74v2 Set | 101788_f_at | - | - |
| Murine Genome U74v2 Set | 102148_f_at | - | - |
| Murine Genome U74v2 Set | 101788_PM_f_at | - | - |
| Murine Genome U74v2 Set | 96419_f_at | - | - |
| Murine Genome U74v2 Set | 102148_PM_f_at | - | - |
| Murine Genome U74v2 Set | 102149_f_at | - | - |
| Murine Genome U74v2 Set | 102149_PM_f_at | - | - |
| Murine Genome U74v2 Set | 96419_PM_f_at | - | - |
| Murine Genome U74v2 Set | 101619_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422406_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1450593_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422404_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422406_at | - | - |
| Mouse Genome 430A 2.0 Array | 1450593_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422403_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422404_PM_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422403_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 456737 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 2970435 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002970435 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2662468 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2662468 | - | - |
| mouse-6 v1.1 (Illumina) | scl0381590.1_8-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2970435 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0381590.1_8-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2970435 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2662468 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2662468 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl0381590.1_8-S | - | - |

C87499
Genomic Map