| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1800000793.mm.1 |
NM_028341.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800000793.mm.1 |
NM_028341.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800000793.mm.1 |
TC1800000105.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1800000796.mm.1 |
NM_028341.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800000796.mm.1 |
NM_028341.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800000796.mm.1 |
TC1800000105.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1800000794.mm.1 |
NM_028341.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800000794.mm.1 |
NM_028341.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800000794.mm.1 |
TC1800000105.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1800000795.mm.1 |
NM_028341.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800000795.mm.1 |
NM_028341.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800000795.mm.1 |
TC1800000105.mm.1 |
MTA TC |