Assay Details
| Gene Symbol: Ttc39c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028341.4 | NP_082617.2 | 4 - 5 | 805 | 74 | |
| GenBank mRNA | AK013267.1 | - | 4 - 5 | 540 | 74 | |
| BC020021.1 | - | 4 - 5 | 555 | 2648772 | 74 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Ttc39c
Genomic Map