Assay Details
| Gene Symbol: Inadl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005784.1 | NP_001005784.1 | 13 - 14 | 1766 | 64 | |
| NM_172696.2 | NP_766284.2 | 13 - 14 | 1766 | 64 | ||
| GenBank mRNA | AK050545.1 | - | 13 - 14 | 1738 | 64 | |
| AK147866.1 | - | 13 - 14 | 1715 | 64 | ||
| AK153597.1 | - | 2 - 3 | 621 | 64 | ||
| AK156166.1 | - | 2 - 3 | 234 | 64 | ||
| BC050846.1 | - | 13 - 14 | 1700 | 64 | ||
| BC062194.1 | - | 1 - 2 | 68 | 64 | ||
| BC082787.1 | - | 2 - 3 | 159 | 64 | ||
| BC141404.1 | - | 13 - 14 | 1742 | 9056017 | 64 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK153597.1 transcripts.
Set Membership: |
|

Inadl
Genomic Map