Assay Details
| Gene Symbol: Inadl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005784.1 | NP_001005784.1 | 14 - 15 | 1824 | 92 | |
| NM_172696.2 | NP_766284.2 | 14 - 15 | 1824 | 92 | ||
| GenBank mRNA | AK050545.1 | - | 14 - 15 | 1796 | 92 | |
| AK147866.1 | - | 14 - 15 | 1773 | 92 | ||
| AK153597.1 | - | 3 - 4 | 679 | 92 | ||
| AK156166.1 | - | 3 - 4 | 292 | 92 | ||
| BC062194.1 | - | 2 - 3 | 126 | 92 | ||
| BC082787.1 | - | 3 - 4 | 217 | 92 | ||
| BC141404.1 | - | 14 - 15 | 1800 | 9056017 | 92 | |
More Information
Set Membership: |
|

Inadl
Genomic Map