Assay Details
| Gene Symbol: Inadl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005784.1 | NP_001005784.1 | 16 - 17 | 2114 | 88 | |
| NM_172696.2 | NP_766284.2 | 16 - 17 | 2114 | 88 | ||
| GenBank mRNA | AK077268.1 | - | 3 - 4 | 396 | 88 | |
| AK147866.1 | - | 16 - 17 | 2063 | 88 | ||
| AK153597.1 | - | 5 - 6 | 969 | 88 | ||
| AK156166.1 | - | 5 - 6 | 582 | 88 | ||
| BB617492.1 | - | 3 - 4 | 393 | 88 | ||
| BC062194.1 | - | 4 - 5 | 416 | 88 | ||
| BC082787.1 | - | 5 - 6 | 507 | 88 | ||
| BC141404.1 | - | 16 - 17 | 2090 | 9056017 | 88 | |
More Information
Updated Mapping Information
Set Membership: |
|

Inadl
Genomic Map