Assay Details
| Gene Symbol: Inadl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005787.1 | NP_001005787.1 | 4 - 5 | 730 | 79 | |
| NM_007704.2 | NP_031730.1 | 4 - 5 | 730 | 79 | ||
| NM_172696.2 | NP_766284.2 | 30 - 31 | 4180 | 79 | ||
| GenBank mRNA | AF060539.1 | - | 4 - 5 | 630 | 79 | |
| AK077268.1 | - | 17 - 18 | 2462 | 79 | ||
| AK147866.1 | - | 30 - 31 | 4129 | 79 | ||
| BC037607.1 | - | 4 - 5 | 402 | 79 | ||
| BC057124.1 | - | 4 - 5 | 409 | 79 | ||
| BC082787.1 | - | 19 - 20 | 2573 | 79 | ||
| BC141404.1 | - | 30 - 31 | 4156 | 9056017 | 79 | |
More Information
Set Membership: |
|

Inadl
Genomic Map