Assay Details
| Gene Symbol: Inadl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005784.1 | NP_001005784.1 | 5 - 6 | 656 | 115 | |
| NM_172696.2 | NP_766284.2 | 5 - 6 | 656 | 115 | ||
| GenBank mRNA | AK050545.1 | - | 5 - 6 | 628 | 115 | |
| AK147866.1 | - | 5 - 6 | 605 | 115 | ||
| BC050846.1 | - | 5 - 6 | 590 | 115 | ||
| BC141404.1 | - | 5 - 6 | 632 | 9056017 | 115 | |
More Information
Set Membership: |
|

Inadl
Genomic Map