| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0400006636.mm.1 |
NM_001005787.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400006636.mm.1 |
NM_001005787.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400006636.mm.1 |
NM_007704.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400006636.mm.1 |
NM_007704.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400006636.mm.1 |
NM_172696.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400006636.mm.1 |
NM_172696.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400006636.mm.1 |
TC0400000982.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400006637.mm.1 |
NM_001005787.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400006637.mm.1 |
NM_001005787.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400006637.mm.1 |
TC0400000982.mm.1 |
MTA TC |