Assay Details
| Gene Symbol: Inadl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005784.1 | NP_001005784.1 | 7 - 8 | 984 | 80 | |
| NM_172696.2 | NP_766284.2 | 7 - 8 | 984 | 80 | ||
| GenBank mRNA | AK050545.1 | - | 7 - 8 | 956 | 80 | |
| AK136048.1 | - | 1 - 2 | 167 | 80 | ||
| AK147866.1 | - | 7 - 8 | 933 | 80 | ||
| BC050846.1 | - | 7 - 8 | 918 | 80 | ||
| BC141404.1 | - | 7 - 8 | 960 | 9056017 | 80 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK136048.1 transcripts.
Set Membership: |
|

Inadl
Genomic Map